siRNA Details
Gene Symbol: PSMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323407.1 | NP_001310336.1 | 3, 4 | 425 | ||
NM_001323408.1 | NP_001310337.1 | 3, 4 | 554 | |||
NM_001323409.1 | NP_001310338.1 | 3, 4 | 554 | |||
NM_001323410.1 | NP_001310339.1 | 3, 4 | 554 | |||
NM_006814.4 | NP_006805.2 | 3, 4 | 554 | |||
NM_178578.3 | NP_848693.2 | 4, 5 | 575 | |||
GenBank mRNA | AK098612.1 | - | 3, 4 | 520 | ||
AK300667.1 | - | 3, 4 | 299 | |||
AK302164.1 | - | 3, 4 | 391 | |||
BC029836.1 | - | 1 | 289 | |||
BC126462.1 | - | 4, 5 | 431 | 8992178 | ||
BI461198.1 | - | 3, 4 | 550 | |||
BI830805.1 | - | 4, 5 | 508 | |||
D88378.1 | - | 3, 4 | 499 | |||
DA949012.1 | - | 3, 4 | 511 | |||
DB028624.1 | - | 4, 5 | 541 | |||
DC398121.1 | - | 3, 4 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
