siRNA Details
Gene Symbol: D1Ertd622e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133825.3 | NP_598586.1 | 3 | 1857 | ||
GenBank mRNA | AK135656.1 | - | 3 | 1807 | ||
AK137265.1 | - | 3 | 2617 | |||
AK152370.1 | - | 3 | 1829 | |||
AK168549.1 | - | 3 | 1832 | |||
BC019375.1 | - | 2 | 1714 | 5008599 | ||
BC023951.1 | - | 3 | 1826 | 5320263 | ||
BC034088.1 | - | 3 | 2321 |
