siRNA Details
Gene Symbol: Miip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170224.1 | - | 6 | 1097 | ||
BC017525.1 | - | 6 | 1047 | 4910951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
