siRNA Details
Gene Symbol: D6Wsu163e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138594.3 | NP_613060.1 | 10 | 1269 | ||
GenBank mRNA | AK019434.1 | - | 2 | 159 | ||
AK037095.1 | - | 10 | 1274 | |||
AK045541.1 | - | 10 | 1270 | |||
AK046707.1 | - | 10 | 1257 | |||
AK137201.1 | - | 10 | 1260 | |||
BC016429.1 | - | 9 | 1217 | 4486831 |
