siRNA Details
Gene Symbol: Adgra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177469.3 | NP_803420.2 | 8 | 3730 | ||
GenBank mRNA | AK032494.1 | - | 1 | 1165 | ||
AK034188.1 | - | 7 | 3293 | |||
BC056493.1 | - | 6 | 3013 | 5704932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
