siRNA Details
Gene Symbol: Tectb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009348.3 | NP_033374.2 | 10 | 2687 | ||
GenBank mRNA | AK010948.1 | - | 4 | 2251 | ||
X99806.2 | - | 10 | 2686 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tectb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009348.3 | NP_033374.2 | 10 | 2687 | ||
GenBank mRNA | AK010948.1 | - | 4 | 2251 | ||
X99806.2 | - | 10 | 2686 |