siRNA Details
Gene Symbol: Tpbpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009411.4 | NP_033437.2 | 5 | 1000 | ||
GenBank mRNA | AK005417.1 | - | 5 | 1001 | ||
AK005454.1 | - | 5 | 1002 | |||
AK011401.1 | - | 5 | 1001 | |||
AK028071.1 | - | 5 | 1000 |
