siRNA Details
Gene Symbol: Tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009429.3 | NP_033455.1 | 6 | 761 | ||
GenBank mRNA | AA269935.1 | - | 2 | 181 | ||
AK150300.1 | - | 6 | 688 | |||
AK150500.1 | - | 6 | 687 | |||
AK150858.1 | - | 6 | 688 | |||
AK151059.1 | - | 6 | 689 | |||
AK151266.1 | - | 6 | 687 | |||
AK151375.1 | - | 6 | 687 | |||
AK151894.1 | - | 6 | 687 | |||
AK152690.1 | - | 6 | 687 | |||
AK152808.1 | - | 6 | 688 | |||
AK159516.1 | - | 6 | 687 | |||
AK159565.1 | - | 6 | 687 | |||
AK165736.1 | - | 6 | 687 | |||
AK166584.1 | - | 6 | 690 | |||
AK167413.1 | - | 6 | 687 | |||
AK167936.1 | - | 6 | 687 | |||
AK168151.1 | - | 6 | 687 | |||
AK169730.1 | - | 6 | 689 | |||
AK198939.1 | - | 3 | 250 | |||
AK204488.1 | - | 3 | 237 | |||
BC092381.1 | - | 6 | 647 | 6824238 | ||
X06407.1 | - | 6 | 688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
