siRNA Details
Gene Symbol: Ubr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009461.2 | NP_033487.2 | 4, 5 | 635 | ||
GenBank mRNA | AF061555.1 | - | 4, 5 | 636 | ||
AK089616.1 | - | 4, 5 | 535 | |||
AK132284.1 | - | 4, 5 | 549 | |||
AK162013.1 | - | 4, 5 | 536 | |||
AK166885.1 | - | 4, 5 | 533 | |||
BC090969.1 | - | 4, 5 | 523 | |||
BC100461.1 | - | 4, 5 | 542 |
