siRNA Details
Gene Symbol: Uros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA272198.1 | - | 4, 5 | 424 | ||
AK135314.1 | - | 5, 6 | 480 | |||
AK145902.1 | - | 5, 6 | 530 | |||
AK148116.1 | - | 5, 6 | 667 | |||
AK164297.1 | - | 5, 6 | 445 | |||
BC068161.1 | - | 5, 6 | 473 | 30358504 | ||
U04439.1 | - | 4, 5 | 394 | |||
U16216.1 | - | 5, 6 | 535 | |||
U18867.2 | - | 5, 6 | 539 |

More Information
Other Species Targeted

