siRNA Details
Gene Symbol: Vav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009500.1 | NP_033526.1 | 13 | 1580 | ||
GenBank mRNA | BC053060.1 | - | 13 | 1557 | 5704852 | |
U37017.1 | - | 13 | 1580 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009500.1 | NP_033526.1 | 13 | 1580 | ||
GenBank mRNA | BC053060.1 | - | 13 | 1557 | 5704852 | |
U37017.1 | - | 13 | 1580 |