siRNA Details
Gene Symbol: ERBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001982.3 | NP_001973.2 | 28 | 4793 | ||
GenBank mRNA | AK124710.1 | - | 11 | 2975 | ||
AK125028.1 | - | 6 | 2335 | |||
BC002706.2 | - | 11 | 2464 | |||
BC041579.1 | - | 13 | 2438 | |||
BC082992.1 | - | 28 | 4632 | 6147464 | ||
M29366.1 | - | 28 | 4616 | |||
M34309.1 | - | 28 | 4715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
