siRNA Details
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025250.3 | NP_001020421.2 | 7 | 1551 | ||
NM_001110266.1 | NP_001103736.1 | 4 | 861 | |||
NM_001110267.1 | NP_001103737.1 | 3 | 789 | |||
NM_001287056.1 | NP_001273985.1 | 7 | 1551 | |||
NM_001287057.1 | NP_001273986.1 | 6 | 1479 | |||
NM_009505.4 | NP_033531.3 | 6 | 1479 | |||
GenBank mRNA | AW913188.1 | - | 2 | 109 | ||
BC061468.1 | - | 6 | 852 | 6816435 | ||
BQ554097.1 | - | 4 | 506 | |||
HQ712122.1 | - | 1 | 46 | |||
M95200.1 | - | 6 | 544 | |||
S37052.1 | - | 7 | 542 | |||
S38083.1 | - | 6 | 470 |

More Information
Other Species Targeted

Gene Ontology Categories:
