siRNA Details
Gene Symbol: Xrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009532.4 | NP_033558.3 | 4 | 491 | ||
GenBank mRNA | AK155376.1 | - | 4 | 468 | ||
AK165569.1 | - | 4 | 446 | |||
AK168334.1 | - | 4 | 441 | |||
AK168727.1 | - | 4 | 448 | |||
BC055900.1 | - | 4 | 467 | 4207343 | ||
U02887.1 | - | 4 | 455 |

More Information
Other Species Targeted

Gene Ontology Categories:
