siRNA Details
Gene Symbol: Zfp101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009542.2 | NP_033568.2 | 4 | 674 | ||
GenBank mRNA | AK076355.1 | - | 4 | 678 | ||
AK076461.1 | - | 1 | 464 | |||
AK078458.1 | - | 4 | 662 | |||
AK169234.1 | - | 4 | 675 | |||
U07861.1 | - | 4 | 634 |
