siRNA Details
Gene Symbol: Zbtb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009547.2 | NP_033573.1 | 3 | 3141 | ||
GenBank mRNA | AK037725.1 | - | 3 | 3262 | ||
AK080769.1 | - | 1 | 1212 | |||
AK082051.1 | - | 3 | 3283 | |||
AK136762.1 | - | 3237 | ||||
AK145517.1 | - | 3 | 2974 | |||
AK146201.1 | - | 3 | 2943 | |||
BC052017.1 | - | 3 | 3141 | 5697838 |
