siRNA Details
Gene Symbol: SERINC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006811.3 | NP_006802.1 | 10 | 1734 | ||
XM_017027601.1 | XP_016883090.1 | 10 | 1628 | |||
GenBank mRNA | AF112227.1 | - | 10 | 1646 | ||
AF153979.1 | - | 10 | 1655 | |||
AK222923.1 | - | 10 | 1656 | |||
AK295120.1 | - | 11 | 1609 | |||
AK309993.1 | - | 11 | 1793 | |||
U49188.1 | - | 10 | 1667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
