siRNA Details
Gene Symbol: Zic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028305.1 | - | 1 | 131 | ||
AK035908.1 | - | 1 | 132 | |||
D78174.1 | - | 1 | 581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
