siRNA Details
Gene Symbol: Tgtp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145164.1 | NP_001138636.1 | 2 | 591 | ||
GenBank mRNA | AK088858.1 | - | 2 | 585 | ||
AK172580.1 | - | 2 | 584 | |||
L38444.1 | - | 2 | 422 |
Gene Symbol: Tgtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011579.3 | NP_035709.3 | 2 | 584 | ||
GenBank mRNA | AK089836.1 | - | 2 | 586 | ||
AK163978.1 | - | 2 | 585 | |||
AK172473.1 | - | 2 | 584 | |||
BC034256.1 | - | 2 | 549 | 3662733 | ||
BC085259.1 | - | 555 | 5389279 | |||
BC093522.1 | - | 2 | 566 | 3988138 | ||
CT010183.1 | - | 1 | 323 | |||
U15636.1 | - | 2 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
