siRNA Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011650.3 | NP_035780.1 | 6 | 1938 | ||
GenBank mRNA | AF234179.1 | - | 6 | 1936 | ||
AK011658.1 | - | 1 | 661 | |||
AK165598.1 | - | 6 | 1982 |
