siRNA Details
Gene Symbol: STON1-GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198593.1 | NP_001185522.1 | 2 | 216 | ||
NM_001198594.1 | NP_001185523.1 | 1 | 152 | |||
NM_172311.2 | NP_758515.1 | 2 | 219 | |||
GenBank mRNA | AF026169.1 | - | 2 | 219 | ||
AK309740.1 | - | 2 | 216 | |||
BC101772.1 | - | 1 | 132 | 8069278 | ||
BC101774.1 | - | 1 | 132 | 8069280 | ||
BC143520.1 | - | 1 | 132 | 9052029 |
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198595.1 | NP_001185524.1 | 3 | 317 | ||
NM_006873.3 | NP_006864.2 | 2 | 200 | |||
GenBank mRNA | AF255310.1 | - | 1 | 115 | ||
AK314464.1 | - | 2 | 200 | |||
AY026319.1 | - | 1 | 105 | |||
BC121063.2 | - | 1 | 115 | 40118538 | ||
BC121064.2 | - | 1 | 115 | 40118539 | ||
CR936807.1 | - | 3 | 300 | |||
DB288942.1 | - | 2 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
