siRNA Details
Gene Symbol: ELF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276457.1 | NP_001263386.1 | 2 | 387 | ||
NM_006874.3 | NP_006865.1 | 2 | 387 | |||
XM_005262803.2 | XP_005262860.1 | 5 | 846 | |||
XM_006714128.2 | XP_006714191.1 | 5 | 846 | |||
XM_006714129.3 | XP_006714192.1 | 4 | 625 | |||
XM_011531710.2 | XP_011530012.1 | 5 | 803 | |||
XM_011531711.2 | XP_011530013.1 | 3 | 352 | |||
XM_011531712.2 | XP_011530014.1 | 2 | 343 | |||
XM_017007858.1 | XP_016863347.1 | 4 | 681 | |||
GenBank mRNA | BC034951.1 | - | 2 | 471 | 4821608 | |
BC065025.1 | - | 2 | 316 | 6044351 | ||
U43189.1 | - | 2 | 284 |

More Information
Other Species Targeted

Gene Ontology Categories:
