siRNA Details
Gene Symbol: Zfp59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011762.3 | NP_035892.2 | 5 | 1792 | ||
GenBank mRNA | AK142951.1 | - | 5 | 1769 | ||
M28514.1 | - | 1 | 1046 |
Gene Symbol: Gm7452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127053.1 | - | 1 | 1568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
