siRNA Details
Gene Symbol: Zfp97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011765.5 | NP_035895.3 | 4 | 1121 | ||
GenBank mRNA | L20450.1 | - | 1089 |
Gene Symbol: Zfp960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005358.2 | NP_001005358.2 | 4 | 1070 | ||
NM_001163919.1 | NP_001157391.1 | 4 | 1070 | |||
GenBank mRNA | AK145357.1 | - | 4 | 1058 | ||
BC018101.1 | - | 4 | 1060 | 3494875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
