siRNA Details
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011767.2 | NP_035897.2 | 13 | 2258 | ||
GenBank mRNA | AF071059.1 | - | 13 | 2256 | ||
AK034963.1 | - | 6 | 827 | |||
AK158938.1 | - | 5 | 566 | |||
BC006962.1 | - | 3 | 305 | |||
BC094220.1 | - | 12 | 2145 | |||
BC141139.1 | - | 13 | 2242 | 9055751 |

More Information
Other Species Targeted

Gene Ontology Categories:
