siRNA Details
Gene Symbol: HNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006895.2 | NP_008826.1 | 2, 3 | 446 | ||
XM_017003948.1 | XP_016859437.1 | 3, 4 | 906 | |||
XM_017003949.1 | XP_016859438.1 | 2, 3 | 279 | |||
GenBank mRNA | AF523358.1 | - | 2, 3 | 373 | ||
AF523359.1 | - | 2, 3 | 373 | |||
AF523360.1 | - | 2, 3 | 373 | |||
AK301462.1 | - | 2, 3 | 204 | |||
AK313804.1 | - | 2, 3 | 204 | |||
BC020677.1 | - | 2, 3 | 205 | 4767331 | ||
D16224.1 | - | 2, 3 | 202 | |||
U08092.1 | - | 2, 3 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
