siRNA Details
Gene Symbol: Adam23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177600.1 | NP_001171071.1 | 8, 9 | 1157 | ||
NM_011780.2 | NP_035910.1 | 8, 9 | 1157 | |||
GenBank mRNA | AB009673.1 | - | 8, 9 | 929 | ||
AK029301.1 | - | 6, 7 | 472 | |||
AK034022.1 | - | 8, 9 | 1152 | |||
AK046677.1 | - | 8, 9 | 1171 |
