siRNA Details
Gene Symbol: Adam20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009548.2 | NP_001009548.1 | 2 | 465 | ||
GenBank mRNA | AK015933.1 | - | 2 | 474 | ||
AK133113.1 | - | 2 | 466 | |||
BC150793.1 | - | 351 | 9087704 |
Gene Symbol: Adam25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011781.2 | NP_035911.2 | 2 | 468 | ||
GenBank mRNA | AF167403.1 | - | 468 | |||
AK016581.1 | - | 2 | 466 |
Gene Symbol: Adam39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025380.3 | NP_001020551.3 | 2 | 478 | ||
GenBank mRNA | AK006820.1 | - | 2 | 440 |

More Information
Other Species Targeted

Gene Ontology Categories:
