siRNA Details
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278282.1 | NP_001265211.1 | 4, 5 | 452 | ||
NM_001278283.1 | NP_001265212.1 | 5, 6 | 532 | |||
NM_001318926.1 | NP_001305855.1 | 4, 5 | 501 | |||
NM_001318928.1 | NP_001305857.1 | 4, 5 | 501 | |||
NM_006916.2 | NP_008847.1 | 3, 4 | 421 | |||
GenBank mRNA | AK056028.1 | - | 3, 4 | 358 | ||
AK093658.1 | - | 5, 6 | 463 | |||
AK096199.1 | - | 4, 5 | 388 | |||
AY947714.1 | - | 2, 3 | 120 | |||
BX403091.2 | - | 4, 5 | 407 | |||
DA592494.1 | - | 4, 5 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
