siRNA Details
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127367.1 | NP_001120839.1 | 8 | 1164 | ||
NM_011847.4 | NP_035977.2 | 8 | 1167 | |||
GenBank mRNA | AB028854.1 | - | 8 | 1069 | ||
AF035962.1 | - | 8 | 1067 | |||
AK133805.1 | - | 8 | 1084 | |||
AK134387.1 | - | 8 | 1126 | |||
AK144182.1 | - | 8 | 1088 | |||
AK151219.1 | - | 9 | 1074 | |||
AK151752.1 | - | 8 | 1041 | |||
AK152626.1 | - | 8 | 1081 | |||
AK152632.1 | - | 8 | 1041 | |||
AK169332.1 | - | 8 | 1073 | |||
AK169767.1 | - | 8 | 1035 | |||
BC003702.1 | - | 8 | 1049 | 3601332 | ||
BC083349.1 | - | 8 | 1059 | 30541723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
