siRNA Details
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005849.1 | NP_001005849.1 | 2 | 284 | ||
NM_006937.3 | NP_008868.3 | 2 | 284 | |||
GenBank mRNA | AK024823.1 | - | 2 | 185 | ||
AK225090.1 | - | 2 | 195 | |||
AK225263.1 | - | 2 | 196 | |||
AK311837.1 | - | 2 | 230 | |||
BC008450.1 | - | 2 | 251 | 4133768 | ||
BC016775.1 | - | 2 | 265 | 4096440 | ||
BC022340.1 | - | 2 | 235 | 4134531 | ||
BC062713.1 | - | 2 | 198 | 30404344 | ||
BC068465.1 | - | 2 | 250 | 5277638 | ||
BC070159.1 | - | 2 | 258 | 6592382 | ||
BC071645.1 | - | 2 | 191 | 3884132 | ||
BC071646.1 | - | 2 | 227 | 4369354 | ||
BC107853.1 | - | 2 | 188 | 5772748 | ||
BF700114.1 | - | 2 | 251 | |||
BG778261.1 | - | 2 | 285 | |||
L76416.1 | - | 2 | 129 | |||
X99585.1 | - | 2 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
