siRNA Details
Gene Symbol: ZNF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029976.2 | NP_001025147.2 | 4 | 404 | ||
NM_006958.2 | NP_008889.2 | 3 | 318 | |||
XM_005272341.2 | XP_005272398.1 | 3 | 295 | |||
XM_005272342.2 | XP_005272399.1 | 3 | 403 | |||
XM_011517298.1 | XP_011515600.1 | 3 | 420 | |||
GenBank mRNA | AF244088.1 | - | 2 | 229 | ||
AK127625.1 | - | 3 | 318 | |||
AK226016.1 | - | 3 | 288 | |||
AL834451.1 | - | 2 | 235 | |||
BC010996.2 | - | 3 | 252 | 3949487 | ||
DQ117529.1 | - | 4 | 404 |
