siRNA Details
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039075.2 | NP_001034164.1 | 9, 10 | 879 | ||
NM_001039076.2 | NP_001034165.1 | 7, 8 | 996 | |||
GenBank mRNA | AF114379.2 | - | 9, 10 | 843 | ||
AJ005621.1 | - | 9, 10 | 872 | |||
AK004020.1 | - | 9, 10 | 887 | |||
AK137181.1 | - | 7, 8 | 979 | |||
AY206011.1 | - | 6, 7 | 913 | |||
BC099596.1 | - | 9, 10 | 846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
