siRNA Details
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039075.2 | NP_001034164.1 | 10 | 956 | ||
NM_001039076.2 | NP_001034165.1 | 8 | 1073 | |||
GenBank mRNA | AF114379.2 | - | 10 | 920 | ||
AJ005621.1 | - | 10 | 949 | |||
AK004020.1 | - | 10 | 964 | |||
AK137181.1 | - | 8 | 1056 | |||
AY206011.1 | - | 7 | 990 | |||
BC099596.1 | - | 10 | 923 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001277158, NM_001277159, NM_001110490, XM_006252645, XM_006252647, and others.
Gene Ontology Categories:
