siRNA Details
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286066.1 | NP_001272995.1 | 16 | 1936 | ||
NM_011938.4 | NP_036068.2 | 16 | 1943 | |||
GenBank mRNA | AF040749.1 | - | 16 | 1813 | ||
CA528447.1 | - | 5 | 542 | |||
CD354984.1 | - | 3 | 283 | |||
Y15797.1 | - | 16 | 1764 | |||
Y15800.1 | - | 16 | 1757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
