siRNA Details
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146153.1 | NP_001139625.1 | 10 | 1931 | ||
NM_011984.2 | NP_036114.2 | 10 | 1922 | |||
GenBank mRNA | AF093261.1 | - | 8 | 1758 | ||
BC005773.1 | - | 10 | 1871 | 3602414 |

More Information
Other Species Targeted

Gene Ontology Categories:
