siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290635.1 | NP_001277564.1 | 4 | 659 | ||
NM_012018.2 | NP_036148.2 | 14 | 2301 | |||
GenBank mRNA | AK088733.1 | - | 4 | 661 | ||
BC050831.1 | - | 6 | 933 | |||
BC060146.1 | - | 4 | 635 | |||
BC079536.1 | - | 2 | 323 | |||
BC158010.1 | - | 13 | 2174 | 9007318 | ||
X79131.1 | - | 6 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
