siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290635.1 | NP_001277564.1 | 6 | 923 | ||
NM_012018.2 | NP_036148.2 | 16 | 2565 | |||
GenBank mRNA | AK088733.1 | - | 6 | 925 | ||
BC050831.1 | - | 8 | 1197 | |||
BC060146.1 | - | 6 | 899 | |||
BC079536.1 | - | 4 | 587 | |||
BC158010.1 | - | 15 | 2438 | 9007318 | ||
X79131.1 | - | 8 | 1222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
