siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270454.1 | NP_001257383.1 | 3 | 221 | ||
NM_001270455.1 | NP_001257384.1 | 3 | 221 | |||
NM_007014.4 | NP_008945.2 | 4 | 274 | |||
XM_011522823.2 | XP_011521125.1 | 5 | 304 | |||
XM_011522825.1 | XP_011521127.1 | 3 | 185 | |||
XM_017022879.1 | XP_016878368.1 | 5 | 378 | |||
XM_017022880.1 | XP_016878369.1 | 5 | 310 | |||
XM_017022881.1 | XP_016878370.1 | 5 | 354 | |||
GenBank mRNA | AJ001982.1 | - | 3 | 202 | ||
AK312792.1 | - | 3 | 201 | |||
BC000108.2 | - | 2 | 123 | |||
BC013645.1 | - | 2 | 108 | 3453212 | ||
BC064531.1 | - | 3 | 160 | 5588092 | ||
DC402912.1 | - | 3 | 221 | |||
JN712744.1 | - | 3 | 191 | |||
U96114.1 | - | 4 | 206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
