siRNA Details
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281742.1 | NP_001268671.1 | 3 | 273 | ||
NM_007019.3 | NP_008950.1 | 3 | 273 | |||
NM_181799.2 | NP_861515.1 | 3 | 273 | |||
NM_181801.3 | NP_861517.1 | 3 | 310 | |||
GenBank mRNA | AK311949.1 | - | 3 | 240 | ||
BC007656.2 | - | 3 | 196 | 3345575 | ||
BC016292.1 | - | 3 | 202 | 3910286 | ||
BC028161.1 | - | 2 | 812 | |||
BC050736.1 | - | 3 | 223 | 6503915 | ||
BF974975.1 | - | 329 | ||||
BI858659.1 | - | 3 | 310 | |||
BM556795.1 | - | 3 | 191 | |||
BM747513.1 | - | 3 | 228 | |||
BP227692.1 | - | 3 | 300 | |||
BT007300.1 | - | 3 | 153 | |||
BT020165.1 | - | 3 | 153 | |||
BT020166.1 | - | 3 | 153 | |||
BU169315.1 | - | 3 | 214 | |||
CN354813.1 | - | 3 | 174 | |||
CR456988.1 | - | 3 | 153 | |||
DB062573.1 | - | 3 | 274 | |||
U73379.1 | - | 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
