siRNA Details
Gene Symbol: Zfp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013705.1 | NP_038733.1 | 5 | 733 | ||
GenBank mRNA | AK014064.1 | - | 4 | 575 | ||
AK032058.1 | - | 1311 | ||||
AK173073.1 | - | 1349 | ||||
AY605122.1 | - | 5 | 733 | |||
BC049120.1 | - | 5 | 734 | 6813126 | ||
BC051972.1 | - | 5 | 933 | 5707652 | ||
Z30174.1 | - | 5 | 733 |
