siRNA Details
Gene Symbol: Magel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013779.2 | NP_038807.4 | 1 | 3984 | ||
GenBank mRNA | AJ243608.1 | - | 1 | 1499 | ||
AK082944.1 | - | 1 | 3957 | |||
AK086725.1 | - | 1 | 3979 | |||
BC054763.1 | - | 1 | 1276 | |||
BC062913.1 | - | 3 | 3489 | |||
BC071222.1 | - | 1 | 1762 |

More Information
Other Species Targeted

Gene Ontology Categories:
