siRNA Details
Gene Symbol: Abcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013790.2 | NP_038818.2 | 30 | 4754 | ||
GenBank mRNA | AB012090.1 | - | 4 | 653 | ||
AB019003.1 | - | 30 | 4705 | |||
AF213387.1 | - | 3 | 632 | |||
BC090629.1 | - | 30 | 4711 | 6839816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
