siRNA Details
Gene Symbol: Mapk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016700.4 | NP_057909.1 | 8 | 987 | ||
GenBank mRNA | AB005663.1 | - | 7 | 820 | ||
AB206575.1 | - | 7 | 762 | |||
AB219977.1 | - | 7 | 796 | |||
AK043102.1 | - | 8 | 987 | |||
AK139498.1 | - | 8 | 975 | |||
BC053027.1 | - | 8 | 957 | 5694771 |
