siRNA Details
Gene Symbol: ADGRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ011001.1 | - | 7, 8 | 1224 | ||
AK075555.1 | - | 7, 8 | 1175 | |||
AK131550.1 | - | 6, 7 | 695 | |||
AK299110.1 | - | 6, 7 | 1064 | |||
BC008770.2 | - | 6, 7 | 1066 | 3139174 | ||
BT007311.1 | - | 5, 6 | 908 | |||
CR936747.1 | - | 6, 7 | 1031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
