siRNA Details
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019634.2 | NP_062608.2 | 8 | 1063 | ||
GenBank mRNA | AK004776.1 | - | 8 | 1023 | ||
AK147306.1 | - | 7 | 6084 | |||
AK147404.1 | - | 6 | 6355 | |||
BC026516.1 | - | 7 | 862 | 4911317 | ||
BC050153.1 | - | 8 | 1011 | |||
BC058603.1 | - | 8 | 1019 | 6529011 | ||
D26483.1 | - | 8 | 1011 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
