siRNA Details
Gene Symbol: Tbl1x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020601.2 | NP_065626.1 | 4 | 282 | ||
GenBank mRNA | AK030547.1 | - | 4 | 497 | ||
AK036996.1 | - | 5 | 361 | |||
AK038674.1 | - | 4 | 283 | |||
BC043105.1 | - | 4 | 405 | 6406579 |
