siRNA Details
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006937.3 | NP_008868.3 | 3 | 344 | ||
GenBank mRNA | AK024823.1 | - | 3 | 245 | ||
AK225090.1 | - | 3 | 255 | |||
AK225263.1 | - | 3 | 256 | |||
AK311837.1 | - | 3 | 290 | |||
BC008450.1 | - | 3 | 311 | 4133768 | ||
BC016775.1 | - | 3 | 325 | 4096440 | ||
BC022340.1 | - | 3 | 295 | 4134531 | ||
BC062713.1 | - | 3 | 258 | 30404344 | ||
BC068465.1 | - | 3 | 310 | 5277638 | ||
BC070159.1 | - | 3 | 318 | 6592382 | ||
BC071645.1 | - | 3 | 251 | 3884132 | ||
BC071646.1 | - | 3 | 287 | 4369354 | ||
BC107853.1 | - | 3 | 248 | 5772748 | ||
BG778261.1 | - | 3 | 345 | |||
L76416.1 | - | 3 | 189 | |||
X99585.1 | - | 3 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
