siRNA Details
Gene Symbol: Tbrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042509.1 | - | 1 | 1096 | ||
AK090121.1 | - | 2419 | ||||
BC095996.1 | - | 1 | 1001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
