siRNA Details
Gene Symbol: Foxg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012560.2 | NP_036692.1 | 1 | 1387 | ||
GenBank mRNA | M87634.1 | - | 1 | 1388 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008241, NM_001160112, BC046958, BC079597, U36760, and others.
Gene Ontology Categories:
